Learn More
Biomatik Corporation Human Transcription Activator Brg1 (Smarca4) Elisa Kit
Supplier:  Biomatik Corporation EKC42640
Encompass_Preferred
Human Transcription Activator Brg1 (Smarca4) Elisa Kit for detecting Swi/Snf Related, Matrix Associated, Actin Dependent Regulator Of Chromatin, Subfamily A, Member 4 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P51532; SMARCA4. Target Synonyms: ATP dependent helicase SMARCA4; ATP-dependent helicase SMARCA4; BAF 190; BAF190; BAF190A; Brahma protein like 1; BRG1; BRG1 associated factor 190A; BRG1 protein; BRG1-associated factor 190A; BRM/SWI2 related gene 1; Global transcription activator homologous sequence; global transcription activator snf2l4; Homeotic gene regulator; hSNF2b; Mitotic growth and transcription activator; MRD16; Nuclear protein GRB1; Protein brahma homolog 1; Protein BRG-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5151