Learn More
Biomatik Corporation Mouse Sortilin(SORT1) ELISA kit
Supplier:  Biomatik Corporation EKC42644
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 6.25 pg/mL-400 pg/mL. Sensitivity: 1.56 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q6PHU5. Target Synonyms: Gp95, NT3, neurotensin receptor 3. Restriction: For Research Use Only.
Catalog No. 50-229-5153