Learn More
Biomatik Corporation Human Serine Protease Inhibitor Kazal-Type 6 (Spink6) Elisa Kit
Supplier:  Biomatik Corporation EKC42647
Encompass_Preferred
Human Serine Protease Inhibitor Kazal-Type 6 (Spink6) Elisa Kit for detecting Serine Peptidase Inhibitor, Kazal Type 6 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6UWN8; SPINK6. Target Synonyms: Acrosin inhibitor; BUSI2; EG433180; ISK6_HUMAN; MGC21394; Protease inhibitor H; Serine protease inhibitor Kazal-type 6; Spink5l1; SPINK6; UNQ844; UNQ844/PRO1782. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5156