Learn More
Biomatik Corporation Human Transcobalamin-2 (Tcn2) Elisa Kit
Supplier:  Biomatik Corporation EKC42656
Encompass_Preferred
Human Transcobalamin-2 (Tcn2) Elisa Kit for detecting Transcobalamin Ii; Macrocytic Anemia in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P20062; TCN2. Target Synonyms: D22S676; D22S750; II; Macrocytic anemia; OTTHUMP00000199117; OTTHUMP00000199118; OTTHUMP00000199119; TC; TC II; TC II deficiency; TC-2; TC2; TCII; TCN2; TCN2 deficiency; TCO2_HUMAN; Transcobalamin 2; Transcobalamin 2 deficiency; Transcobalamin II; Transcobalamin II; macrocytic anemia; Transcobalamin-2; Vitamin B12 binding protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5162