Learn More
Biomatik Corporation Human Thy-1 Membrane Glycoprotein (Thy1) Elisa Kit
Supplier:  Biomatik Corporation EKC42666
Encompass_Preferred
Human Thy-1 Membrane Glycoprotein (Thy1) Elisa Kit for detecting Thy-1 Cell Surface Antigen in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04216; THY1. Target Synonyms: CD7; CD90; CD90 antigen; CDw90; FLJ33325; MGC128895; T25; Theta antigen; Thy 1; Thy 1 cell surface antigen; Thy 1 membrane glycoprotein; Thy 1 T cell antigen; Thy 1.2; Thy-1 antigen; Thy-1 membrane glycoprotein; Thy1; Thy1 antigen; Thy1 T cell antigen; Thy1.1; Thy1.2; THY1_HUMAN; Thymus cell antigen 1; theta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5168