Learn More
Biomatik Corporation Human Tumor Necrosis Factor Receptor Superfamily Member 14 (Tnfrsf14) Elisa Kit
Supplier:  Biomatik Corporation EKC42681
Encompass_Preferred
Human Tumor Necrosis Factor Receptor Superfamily Member 14 (Tnfrsf14) Elisa Kit for detecting Tumor Necrosis Factor Receptor Superfamily, Member 14 (Herpesvirus Entry Mediator) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q92956; TNFRSF14. Target Synonyms: HVEML; ATAR; CD270; CD40 like protein precursor; Herpes virus entry mediator A; Herpesvirus entry mediator A; Herpesvirus entry mediator; Herpesvirus entry mediator ligand; HveA; HVEM; HVEM L; LIGHT; LIGHTR; TNFRSF14; TNFSF 14; TNR14_HUMAN; TR2; Tumor necrosis factor receptor like gene2; Tumor necrosis factor receptor superfamily member 14; Tumor necrosis factor receptor superfamily member 14 precursor; Tumor necrosis factor receptor-like 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5171