Learn More
Biomatik Corporation Human Tumor Necrosis Factor Receptor Superfamily Member 25 (Tnfrsf25) Elisa Kit
Supplier:  Biomatik Corporation EKC42683
Encompass_Preferred
Human Tumor Necrosis Factor Receptor Superfamily Member 25 (Tnfrsf25) Elisa Kit for detecting Tumor Necrosis Factor Receptor Superfamily, Member 25 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.24-15ng/ml. Sensitivity: 0.06ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q93038; TNFRSF25. Target Synonyms: Apo 3; Apo-3; Apo3; Apoptosis inducing receptor AIR; Apoptosis inducing receptor; Apoptosis mediating receptor; Apoptosis mediating receptor DR 3; Apoptosis mediating receptor DR3; Apoptosis mediating receptor TRAMP; Apoptosis-inducing receptor AIR; Apoptosis-mediating receptor DR3; Apoptosis-mediating receptor TRAMP; DDR 3; DDR3; Death domain receptor 3; Death domain receptor 3 soluble form; Death receptor 3; Death receptor beta; DR 3; DR3; LARD; Lymphocyte associated receptor of death. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5173