Learn More
Biomatik Corporation Mouse Tumor necrosis factor ligand superfamily member 14(TNFSF14) ELISA kit
Supplier:  Biomatik Corporation EKC42686
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 25 pg/mL-1600 pg/mL. Sensitivity: 6.25 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9QYH9. Target Synonyms: CD258, HVEML, LIGHT, LTg, TR2, delta transmembrane LIGHT|herpesvirus entry mediator A|ligand for herpesvirus entry mediator|tumor necrosis factor ligand superfamily, member 14|tumor necrosis factor. Restriction: For Research Use Only.
Catalog No. 50-229-5176