Learn More
Biomatik Corporation Mouse Tumor Necrosis Factor Ligand Superfamily Member 4 (Tnfsf4) Elisa Kit
Supplier:  Biomatik Corporation EKC42688
Encompass_Preferred
Mouse Tumor Necrosis Factor Ligand Superfamily Member 4 (Tnfsf4) Elisa Kit for detecting Tumor Necrosis Factor (Ligand) Superfamily, Member 4 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P43488; Tnfsf4. Target Synonyms: Tnfsf4; Ox40l; Txgp1l; Tumor necrosis factor ligand superfamily member 4; OX40 ligand; OX40L; CD antigen CD252. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5178