Learn More
Biomatik Corporation Human Troponin I, fast skeletal muscle(TNNI2) ELISA kit
Supplier:  Biomatik Corporation EKC42691
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P48788. Target Synonyms: AMCD2B, DA2B, FSSV, fsTnI, OTTHUMP00000014141|OTTHUMP00000014142|fast-twitch skeletal muscle troponin I|troponin I fast twitch 2|troponin I, fast-twitch skeletal muscle isoform|troponin I, skeletal,. Restriction: For Research Use Only.
Catalog No. 50-229-5180