Learn More
Biomatik Corporation Human Troponin T, Fast Skeletal Muscle (Tnnt3) Elisa Kit
Supplier:  Biomatik Corporation EKC42694
Encompass_Preferred
Human Troponin T, Fast Skeletal Muscle (Tnnt3) Elisa Kit for detecting Troponin T Type 3 (Skeletal, Fast) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.81-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P45378; TNNT3. Target Synonyms: AMCD2B; Beta TnTF; Beta-TnTF; DA2B; DKFZp779M2348; fast skeletal muscle; Fast skeletal muscle troponin T; FSSV; fTnT; TNNT 3; Tnnt3; TNNT3_HUMAN; TnTf; Troponin T; Troponin T fast skeletal muscle; Troponin T type 3 (skeletal fast); Troponin T type 3; Troponin T3 skeletal fast. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5183