Learn More
Biomatik Corporation Human Urocortin-2 (Ucn2) Elisa Kit
Supplier:  Biomatik Corporation EKC42705
Encompass_Preferred
Human Urocortin-2 (Ucn2) Elisa Kit for detecting Urocortin 2 in serum, plasma, tissue homogenates, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q96RP3; UCN2. Target Synonyms: SRP; Stresscopin related peptide; Stresscopin-related peptide; Ucn II; Ucn2; UCN2_HUMAN; UCNI; UCNII; UR; Urocortin II; Urocortin related peptide; Urocortin-2; Urocortin-related peptide; URP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5190