Learn More
Biomatik Corporation Mouse Protein Wnt-4(WNT4) ELISA kit
Supplier:  Biomatik Corporation EKC42713
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P22724. Target Synonyms: RP1-224A6.7, SERKAL, WNT-4, WNT-4 protein. Restriction: For Research Use Only.
Catalog No. 50-229-5193