Learn More
Biomatik Corporation Human 14-3-3 Protein Gamma (Ywhag) Elisa Kit
Supplier:  Biomatik Corporation EKC42724
Encompass_Preferred
Human 14-3-3 Protein Gamma (Ywhag) Elisa Kit for detecting Tyrosine 3-Monooxygenase/Tryptophan 5-Monooxygenase Activation Protein, Gamma Polypeptide in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P61981; YWHAG. Target Synonyms: 14 3 3 gamma; 14 3 3 protein gamma; 14 3 3 protein gamma subtype; 14 3 3gamma; 14-3-3 protein gamma; 1433G_HUMAN; 3 monooxygenase/tryptophan 5 monooxgenase activation protein gamma polypeptide; KCIP 1; KCIP-1; KCIP1; N-terminally processed; Protein kinase C inhibitor protein 1; Tyrosine 3 monooxygenase/tryptophan 5 monooxygenase activation protein gamma polypeptide; Ywhag. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5199