Learn More
Biomatik Corporation Rat C-C Motif Chemokine 6 (Ccl6) Elisa Kit
Supplier:  Biomatik Corporation EKC42728
Encompass_Preferred
Rat C-C Motif Chemokine 6 (Ccl6) Elisa Kit for detecting C-C Motif Chemokine 6 (Ccl6) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q68FP3; Ccl6. Target Synonyms: Ccl6C-C motif chemokine 6; Small-inducible cytokine A6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5201