Learn More
Biomatik Corporation Pig Hemoglobin (Hb) Elisa Kit
Supplier:  Biomatik Corporation EKC42734
Encompass_Preferred
Pig Hemoglobin (Hb) Elisa Kit for detecting Hemoglobin (Hb) in serum, plasma, lysate for rbc. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.625-40ug/ml. Sensitivity: 0.312ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5206