Learn More
Biomatik Corporation Human Midregional Pro-Atrial Natriuretic Peptide (Mr-Proanp) Elisa Kit
Supplier:  Biomatik Corporation EKC42763
Encompass_Preferred
Human Midregional Pro-Atrial Natriuretic Peptide (Mr-Proanp) Elisa Kit for detecting Midregional Pro-Atrial Natriuretic Peptide (Mr-Proanp) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 39-2500pmol/l. Sensitivity: 9.8pmol/l. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5219