Learn More
Biomatik Corporation Mouse chemokine (C-C motif) ligand 1 ELISA Kit
Supplier:  Biomatik Corporation EKC42816
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 7.8 pg/ml -500 pg/ml. Sensitivity: 1.95 pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Not Available. Assay Time: 1-5 hours. UniProt ID: P10146. Target Synonyms: I-309, P500, SCYA1, SISe, TCA3, T lymphocyte-secreted protein I-309|inflammatory cytokine I-309|small inducible cytokine A1|small inducible cytokine A1 (I-309, homologous to mouse Tca-3). Restriction: For Research Use Only.
Catalog No. 50-229-5232