Learn More
Biomatik Corporation Mouse Tumor necrosis factor receptor superfamily member 18(TNFRSF18) ELISA kit
Supplier:  Biomatik Corporation EKC42822
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 78 pg/ml - 5000 pg/ml. Sensitivity: 19.5 pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Not Available. Assay Time: 1-5 hours. UniProt ID: O35714. Target Synonyms: AITR, GITR, GITR-D, TNF receptor superfamily activation-inducible protein|activation-inducible TNFR family receptor|glucocorticoid-induced TNFR-related protein. Restriction: For Research Use Only.
Catalog No. 50-229-5238