Learn More
Biomatik Corporation Human CP (Ceruloplasmin) ELISA Kit
Supplier:  Biomatik Corporation EKF57201
Encompass_Preferred
Human CP (Ceruloplasmin) ELISA Kit for detecting CP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P00450. Target Synonyms: CP; Ceruloplasmin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5249