Learn More
Biomatik Corporation Human COL28A1 (Collagen alpha-1 (XXVIII) chain) ELISA Kit
Supplier:  Biomatik Corporation EKF57211
Encompass_Preferred
Human COL28A1 (Collagen alpha-1 (XXVIII) chain) ELISA Kit for detecting COL28A1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10pmol/ml. Sensitivity: 0.094pmol/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q2UY09. Target Synonyms: COL28A1; Collagen alpha-1; XXVIII chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5253