Learn More
Biomatik Corporation Human GAD2 (Glutamate decarboxylase 2) ELISA Kit
Supplier:  Biomatik Corporation EKF57214
Encompass_Preferred
Human GAD2 (Glutamate decarboxylase 2) ELISA Kit for detecting GAD2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q05329. Target Synonyms: GAD2; GAD65; GAD-65; glutamate decarboxylase 2; Glutamate decarboxylase 65 kDa isoform; Glutamate decarboxylase-2; pancreas;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5256