Learn More
Biomatik Corporation Human TPSB2 (Tryptase beta-2) ELISA Kit
Supplier:  Biomatik Corporation EKF57233
Encompass_Preferred
Human TPSB2 (Tryptase beta-2) ELISA Kit for detecting TPSB2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P20231. Target Synonyms: TPSB2; Tryptase beta-2; TPS2; Tryptase-C; mast cell tryptase beta II; mast cell tryptase beta III; tryptase beta 2; gene; pseudogene; tryptase beta II; tryptase beta III; Tryptase II; tryptase III; tryptase-2; Tryptase-2; tryptaseB; tryptaseC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5263