Learn More
Biomatik Corporation Human SLC30A8 (Zinc transporter 8) ELISA Kit
Supplier:  Biomatik Corporation EKF57242
Encompass_Preferred
Human SLC30A8 (Zinc transporter 8) ELISA Kit for detecting SLC30A8 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8IWU4. Target Synonyms: ZNT8; SLC30A8; ZnT-8; Solute carrier family 30 member 8; zinc transporter 8; zinc transporter ZnT-8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5267