Learn More
Biomatik Corporation Human LOXL3 (Lysyl oxidase homolog 3) ELISA Kit
Supplier:  Biomatik Corporation EKF57255
Encompass_Preferred
Human LOXL3 (Lysyl oxidase homolog 3) ELISA Kit for detecting LOXL3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.469-30pmol/ml. Sensitivity: 0.281pmol/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P58215. Target Synonyms: LOXL3; Lysyl oxidase homolog 3; LOL3; LOXL; lysyl oxidase-like 3; Lysyl oxidase-like protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5273