Learn More
Biomatik Corporation Human HBA1 (Hemoglobin subunit alpha) ELISA Kit
Supplier:  Biomatik Corporation EKF57263
Encompass_Preferred
Human HBA1 (Hemoglobin subunit alpha) ELISA Kit for detecting HBA1 in red blood cell lysis, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P69905. Target Synonyms: HBA1; Hemoglobin subunit alpha; Alpha-globin; Hemoglobin alpha chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5278