Learn More
Biomatik Corporation Human PGA3 (Pepsin A) ELISA Kit
Supplier:  Biomatik Corporation EKF57271
Encompass_Preferred
Human PGA3 (Pepsin A) ELISA Kit for detecting PGA3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P0DJD8. Target Synonyms: PGA; Pepsinogen A; PGA3; PGA4; pepsin A; pepsinogen 4; group I; pepsinogen A; pepsinogen A4; PGA5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5281