Learn More
Biomatik Corporation Human F13A1 (Coagulation factor XIII A chain) ELISA Kit
Supplier:  Biomatik Corporation EKF57276
Encompass_Preferred
Human F13A1 (Coagulation factor XIII A chain) ELISA Kit for detecting F13A1 in plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P00488. Target Synonyms: F13A1; F13A; ; Coagulation Factor XIII A1 Polypeptide; Transglutaminase A chain; Coagulation factor XIIIa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5285