Learn More
Biomatik Corporation Human MCPIP (Monocyte Chemoattractant Protein-induced Protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57329
Encompass_Preferred
Human MCPIP (Monocyte Chemoattractant Protein-induced Protein 1) ELISA Kit for detecting MCPIP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.906-250pg/ml. Sensitivity: 2.344pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q5D1E8. Target Synonyms: Ribonuclease ZC3H12A; MCP-induced protein 1; Zinc finger CCCH domain-containing protein 12A; MCPIP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5295