Learn More
Biomatik Corporation Human CHIT1 (Chitotriosidase-1) ELISA Kit
Supplier:  Biomatik Corporation EKF57345
Encompass_Preferred
Human CHIT1 (Chitotriosidase-1) ELISA Kit for detecting CHIT1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q13231. Target Synonyms: CHIT1; Chitotriosidase-1; Chitinase-1; CHI3; CHIT; plasma methylumbelliferyl tetra-N-acetylchitotetraoside hydrolase; FLJ00314; MGC125322. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5302