Learn More
Biomatik Corporation Human ASRGL1 (L-asparaginase) ELISA Kit
Supplier:  Biomatik Corporation EKF57350
Encompass_Preferred
Human ASRGL1 (L-asparaginase) ELISA Kit for detecting ASRGL1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q7L266. Target Synonyms: ASRGL1; L-asparaginase; L-asparagine amidohydrolase; Isoaspartyl dipeptidase; Asparaginase-like protein 1; ALP; CRASH. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5305