Learn More
Biomatik Corporation Human PIK3CD (Phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta isoform) ELISA Kit
Supplier:  Biomatik Corporation EKF57353
Encompass_Preferred
Human PIK3CD (Phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta isoform) ELISA Kit for detecting PIK3CD in cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O00329. Target Synonyms: PIK3CD; Phosphatidylinositol-4; 5-bisphosphate 3-kinase catalytic subunit delta isoform; PI3-kinase subunit delta; PI3K-delta; PI3Kdelta; PtdIns-3-kinase subunit delta; Phosphatidylinositol-4; 5-bisphosphate 3-kinase 110 kDa catalytic subunit delta; PtdIns-3-kinase subunit p110-delta; p110delta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5308