Learn More
Biomatik Corporation Human LGALS3BP (Galectin-3-binding protein) ELISA Kit
Supplier:  Biomatik Corporation EKF57376
Encompass_Preferred
Human LGALS3BP (Galectin-3-binding protein) ELISA Kit for detecting LGALS3BP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.75ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q08380. Target Synonyms: LGALS3BP; Galectin-3-binding protein; Tumor-associated antigen 90K; Mac-2-binding protein; MAC2BP; Mac-2 BP; Basement membrane autoantigen p105; Lectin galactoside-binding soluble 3-binding protein; M2BP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5315