Learn More
Biomatik Corporation Human GREM2 (Gremlin-2) ELISA Kit
Supplier:  Biomatik Corporation EKF57382
Encompass_Preferred
Human GREM2 (Gremlin-2) ELISA Kit for detecting GREM2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9H772. Target Synonyms: GREM2; Gremlin-2; Protein related to DAN and cerberus; Cysteine knot superfamily 1; BMP antagonist 2; DAN domain family member 3; CKTSF1B2; DAND3; PRDC; Protein related to DAN and cerberus; gremlin 2; cysteine knot superfamily; homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5319