Learn More
Biomatik Corporation Human LTBP2 (Latent-transforming growth factor beta-binding protein 2) ELISA Kit
Supplier:  Biomatik Corporation EKF57388
Encompass_Preferred
Human LTBP2 (Latent-transforming growth factor beta-binding protein 2) ELISA Kit for detecting LTBP2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q14767. Target Synonyms: LTBP2; LTBP-2; C14orf141; GLC3D; latent transforming growth factor beta binding protein 2; LTBP3; chromosome 14 open reading frame 141; Latent TGF-beta bp2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5323