Learn More
Biomatik Corporation Human IL32 (Interleukin-32) ELISA Kit
Supplier:  Biomatik Corporation EKF57396
Encompass_Preferred
Human IL32 (Interleukin-32) ELISA Kit for detecting IL32 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P24001. Target Synonyms: IL-32; Interleukin 32; NK4; TAIF; IL-32; IL-32alpha; IL-32beta; interleukin-32; interleukin-32 theta; natural killer cell transcript 4; Natural killer cells protein 4; NK4IL-32delta; TAIFa; TAIFb; TAIFc; TAIFd; TAIFIL-32gamma; Tumor necrosis factor alpha-inducing factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5327