Learn More
Biomatik Corporation Human IL4R (Interleukin-4 receptor) ELISA Kit
Supplier:  Biomatik Corporation EKF57430
Encompass_Preferred
Human IL4R (Interleukin-4 receptor) ELISA Kit for detecting IL4R in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P24394. Target Synonyms: IL4R; Interleukin-4 receptor subunit alpha; IL-4 receptor subunit alpha; IL-4R subunit alpha; IL-4R-alpha; IL-4RA; IL4RA; interleukin 4 receptor; interleukin-4 receptor alpha chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5345