Learn More
Biomatik Corporation Human PIINP (N-terminal propeptide of Collagen alpha-1 (II) chain) ELISA Kit
Supplier:  Biomatik Corporation EKF57460
Encompass_Preferred
Human PIINP (N-terminal propeptide of Collagen alpha-1 (II) chain) ELISA Kit for detecting PIINP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 93.75-6000pg/ml. Sensitivity: 56.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02458. Target Synonyms: PIINP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5358