Learn More
Biomatik Corporation Human SOD3 (Extracellular superoxide dismutase [Cu-Zn]) ELISA Kit
Supplier:  Biomatik Corporation EKF57465
Encompass_Preferred
Human SOD3 (Extracellular superoxide dismutase [Cu-Zn]) ELISA Kit for detecting SOD3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08294. Target Synonyms: SOD3; Extracellular superoxide dismutase [Cu-Zn]; EC-SOD; extracellular superoxide dismutase [Cu-Zn]; superoxide dismutase 3; extracellular. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5360