Learn More
Biomatik Corporation Human TREML1 (Trem-like transcript 1 protein) ELISA Kit
Supplier:  Biomatik Corporation EKF57476
Encompass_Preferred
Human TREML1 (Trem-like transcript 1 protein) ELISA Kit for detecting TREML1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q86YW5. Target Synonyms: TREML1; Trem-like transcript 1 protein; Triggering receptor expressed on myeloid cells-like protein 1; TLT-1; TLT1; trem-like transcript 1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5365