Learn More
Biomatik Corporation Human SEMA3A (Semaphorin-3A) ELISA Kit
Supplier:  Biomatik Corporation EKF57479
Encompass_Preferred
Human SEMA3A (Semaphorin-3A) ELISA Kit for detecting SEMA3A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q14563. Target Synonyms: SEMA3A; Sema3A; ; semaphorin 3A; coll-1; collapsin 1; Hsema-I; Hsema-III; sema domain; immunoglobulin domain; Ig; short basic domain; secreted; Sema III; SEMA1; SEMAD; SEMAIII; SEMAL; semaphorin D; Semaphorin III; semaphorin L; semaphorin-3A; SemD; Semaphorin III; Sema III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5368