Learn More
Biomatik Corporation Human TEC (Tyrosine-protein kinase Tec) ELISA Kit
Supplier:  Biomatik Corporation EKF57492
Encompass_Preferred
Human TEC (Tyrosine-protein kinase Tec) ELISA Kit for detecting TEC in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P42680. Target Synonyms: TEC; PSCTK4; EC 2.7.10; EC 2.7.10.2; MGC126760; MGC126762; tyrosine-protein kinase Tec; tec protein tyrosine kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5375