Learn More
Biomatik Corporation Human ADIPOR2 (Adiponectin Receptor 2) ELISA Kit
Supplier:  Biomatik Corporation EKF57528
Encompass_Preferred
Human ADIPOR2 (Adiponectin Receptor 2) ELISA Kit for detecting ADIPOR2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q86V24. Target Synonyms: ADIPOR2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5393