Learn More
Biomatik Corporation Human AP13 (Apelin 13) ELISA Kit
Supplier:  Biomatik Corporation EKF57539
Encompass_Preferred
Human AP13 (Apelin 13) ELISA Kit for detecting AP13 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 75pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: Q9ULZ1. Target Synonyms: AP13; Apelin 13. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5398