Learn More
Biomatik Corporation Human CA6 (Carbonic Anhydrase VI) ELISA Kit
Supplier:  Biomatik Corporation EKF57560
Encompass_Preferred
Human CA6 (Carbonic Anhydrase VI) ELISA Kit for detecting CA6 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1-64ng/ml. Sensitivity: 0.602ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antibody. Assay Time: 3h, 30 min. Accession Number: P23280. Target Synonyms: CA6; CA-VI; CAVI; Gustin; Gustin; Carbonate dehydRatase VI; carbonic anhydrase 6; carbonic anhydrase VIGUSTIN; CA-VI; EC 4.2.1.1; MGC21256; Salivary carbonic anhydrase; Secreted carbonic anhydrase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5405