Learn More
Biomatik Corporation Human Scgb1a1 (Uteroglobin) ELISA Kit
Supplier:  Biomatik Corporation EKF57564
Encompass_Preferred
Human Scgb1a1 (Uteroglobin) ELISA Kit for detecting Scgb1a1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P11684. Target Synonyms: CC16; Blastokinin; CC10; CCPBP; CCSP; Clara; PCB-BP; SCGB1A1; UGB; blastokinin; CC10Urine protein 1; CC16; CCSPCCPBP; Clara cell phospholipid-binding protein; Clara cells 10 kDa secretory protein; Secretoglobin family 1A member 1; secretoglobin; family 1A; member 1; uteroglobin; UGBUP-1; UP1; Urinary protein 1; uteroglobin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5407