Learn More
Biomatik Corporation Human COL2 (Collagen Type Ⅱ) ELISA Kit
Supplier:  Biomatik Corporation EKF57573
Encompass_Preferred
Human COL2 (Collagen Type Ⅱ) ELISA Kit for detecting COL2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 46.875-3000pg/ml. Sensitivity: 28.125pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02458. Target Synonyms: COL2; Collagen Type Ⅱ; Chondrocalcin; COL2A1; SEDC; Alpha-1 type II collagen; ANFH; AOM; cartilage collagen; COL11A3; Collagen 2; collagen alpha-1; II chain; collagen II; alpha-1 polypeptide; collagen type 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5409