Learn More
Biomatik Corporation Human HBβ (Hemoglobin Beta) ELISA Kit
Supplier:  Biomatik Corporation EKF57632
Encompass_Preferred
Human HBβ (Hemoglobin Beta) ELISA Kit for detecting HBβ in red blood cell lysis, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P68871. Target Synonyms: HBβ; HBB; Hemoglobin subunit beta; Beta-globin; Hemoglobin beta chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5429