Learn More
Biomatik Corporation Human MUC13 (Mucin 13) ELISA Kit
Supplier:  Biomatik Corporation EKF57658
Encompass_Preferred
Human MUC13 (Mucin 13) ELISA Kit for detecting MUC13 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9H3R2. Target Synonyms: MUC13; Mucin 13. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5437