Learn More
Biomatik Corporation Human OFQ/N (Orphanin FQ/Nociceptin) ELISA Kit
Supplier:  Biomatik Corporation EKF57670
Encompass_Preferred
Human OFQ/N (Orphanin FQ/Nociceptin) ELISA Kit for detecting OFQ; N in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 4.688-300pg/ml. Sensitivity: 2.813pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q13519. Target Synonyms: OFQ; N. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5445